Search
Search
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007709.1 | NP_001007710.1 | 5 - 6 | 1071 | 101 | |
| XM_006242081.2 | XP_006242143.1 | 5 - 6 | 1124 | 101 | ||
| XM_006242083.2 | XP_006242145.1 | 6 - 7 | 2130 | 101 | ||
| XM_008765717.1 | XP_008763939.1 | 5 - 6 | 1257 | 101 | ||
| XM_008765718.1 | XP_008763940.1 | 6 - 7 | 2884 | 101 | ||
| GenBank mRNA | BC083593.1 | - | 5 - 6 | 1071 | 7132737 | 101 |
Set Membership: |
|