Search
Search
| Gene Symbol: Zfat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134957.1 | NP_001128429.1 | 12 - 13 | 3252 | 65 | |
| XM_006241712.2 | XP_006241774.1 | 13 - 14 | 5429 | 65 | ||
| XM_006241713.2 | XP_006241775.1 | 12 - 13 | 5405 | 65 | ||
| XM_006241714.2 | XP_006241776.1 | 13 - 14 | 5429 | 65 | ||
| XM_008765515.1 | XP_008763737.1 | 13 - 14 | 3407 | 65 | ||
Set Membership: |
|