Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01503296_m1
          See other TMCC3 GE Assays ›
          Gene Symbol:
          Tmcc3
          Gene Name:
          transmembrane and coiled-coil domain family 3
          Gene Aliases:
          RGD1307241
          Chromosome Location:
          Chr.7: 35472102 - 35543768 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01503296_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmcc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108084.1 NP_001101554.1 3 - 4 1310 69
          XM_006241255.2 XP_006241317.1 3 - 4 1290 69
          XM_006241261.2 XP_006241323.1 4 - 5 1485 69
          XM_006241263.2 XP_006241325.1 3 - 4 2133 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000529.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P397423 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P227510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P227510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P397423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P227510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P397423 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P397423 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P227510 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P227510 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895002 - -
          Affymetrix Rat Gene 1.0 ST Array 10895001 - -
          Affymetrix Rat Gene 1.0 ST Array 10895000 - -
          Affymetrix Rat Gene 1.0 ST Array 10895003 - -
          Affymetrix Rat Gene 1.1 ST Array 10894999 - -
          Rat Genome 230 2.0 Array 1390832_at - -
          Rat Genome 230 2.0 Array 1393822_at - -
          Rat Genome 230 2.0 Array 1393822_PM_at - -
          Rat Genome 230 2.0 Array 1390832_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700004462.rn.1 TC0700000529.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004463.rn.1 TC0700000529.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004464.rn.1 TC0700000529.rn.1 RTA TC
          Rat Expression Set 230 1390832_at - -
          Rat Expression Set 230 1393822_PM_at - -
          Rat Expression Set 230 1393822_at - -
          Rat Expression Set 230 1390832_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894999 - -
          Rat Genome U34 Set rc_AA925789_at - -
          Rat Genome U34 Set rc_AI059241_at - -
          Rat Genome U34 Set rc_AI044267_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21832288 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355918 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline