Search
Search
| Gene Symbol: E2f7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108092.1 | NP_001101562.1 | 2 - 3 | 421 | 64 | |
| XM_006241320.2 | XP_006241382.1 | 2 - 3 | 312 | 64 | ||
| XM_006241321.2 | XP_006241383.1 | 2 - 3 | 154 | 64 | ||
Set Membership: |
|