Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01503718_m1
          See other ALDH1L2 GE Assays ›
          Gene Symbol:
          Aldh1l2
          Gene Name:
          aldehyde dehydrogenase 1 family, member L2
          Gene Aliases:
          RGD1309458
          Chromosome Location:
          Chr.7: 26375866 - 26425838 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01503718_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh1l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191778.1 NP_001178707.1 18 - 19 2407 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000417.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          biosynthetic process
          10-formyltetrahydrofolate catabolic process
          methylation
          oxidation-reduction process
          aldehyde dehydrogenase (NAD) activity
          methyltransferase activity
          formyltetrahydrofolate dehydrogenase activity
          hydroxymethyl-, formyl- and related transferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P506064 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P506064 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P506064 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P506064 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P506064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894584 - -
          Affymetrix Rat Gene 1.0 ST Array 10894603 - -
          Affymetrix Rat Gene 1.0 ST Array 10894588 - -
          Affymetrix Rat Gene 1.0 ST Array 10894605 - -
          Affymetrix Rat Gene 1.0 ST Array 10894583 - -
          Affymetrix Rat Gene 1.0 ST Array 10894589 - -
          Affymetrix Rat Gene 1.0 ST Array 10894600 - -
          Affymetrix Rat Gene 1.0 ST Array 10894587 - -
          Affymetrix Rat Gene 1.0 ST Array 10894601 - -
          Affymetrix Rat Gene 1.0 ST Array 10894596 - -
          Affymetrix Rat Gene 1.0 ST Array 10894592 - -
          Affymetrix Rat Gene 1.0 ST Array 10894591 - -
          Affymetrix Rat Gene 1.0 ST Array 10894585 - -
          Affymetrix Rat Gene 1.0 ST Array 10894593 - -
          Affymetrix Rat Gene 1.0 ST Array 10894586 - -
          Affymetrix Rat Gene 1.0 ST Array 10894599 - -
          Affymetrix Rat Gene 1.0 ST Array 10894595 - -
          Affymetrix Rat Gene 1.0 ST Array 10894594 - -
          Affymetrix Rat Gene 1.0 ST Array 10894590 - -
          Affymetrix Rat Gene 1.0 ST Array 10894602 - -
          Affymetrix Rat Gene 1.0 ST Array 10894598 - -
          Affymetrix Rat Gene 1.0 ST Array 10894597 - -
          Affymetrix Rat Gene 1.0 ST Array 10894604 - -
          Affymetrix Rat Clariom S Assay PSR0700003573.rn.1 TC0700000417.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700003570.rn.1 TC0700000417.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700003572.rn.1 TC0700000417.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10894582 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352945 - -
          RatRef-12 Expression BeadChip ILMN_64975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline