Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01503726_m1
          See other NAV3 GE Assays ›
          Gene Symbol:
          Nav3
          Gene Name:
          neuron navigator 3
          Gene Aliases:
          RGD1306259
          Chromosome Location:
          Chr.7: 52164805 - 52508145 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01503726_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nav3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191782.1 NP_001178711.1 12 - 13 2838 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002667.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P190136 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P190136 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P190136 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P190136 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P246241 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P190136 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10902276 - -
          Affymetrix Rat Gene 1.0 ST Array 10902285 - -
          Affymetrix Rat Gene 1.0 ST Array 10902289 - -
          Affymetrix Rat Gene 1.0 ST Array 10902281 - -
          Affymetrix Rat Gene 1.0 ST Array 10902278 - -
          Affymetrix Rat Gene 1.0 ST Array 10902286 - -
          Affymetrix Rat Gene 1.0 ST Array 10902283 - -
          Affymetrix Rat Gene 1.0 ST Array 10902287 - -
          Affymetrix Rat Gene 1.0 ST Array 10902291 - -
          Affymetrix Rat Gene 1.0 ST Array 10902277 - -
          Affymetrix Rat Gene 1.0 ST Array 10902275 - -
          Affymetrix Rat Gene 1.0 ST Array 10902288 - -
          Affymetrix Rat Gene 1.0 ST Array 10902284 - -
          Affymetrix Rat Gene 1.1 ST Array 10902249 - -
          Affymetrix Rat Gene 1.1 ST Array 10902280 - -
          Affymetrix Rat Gene 1.1 ST Array 10902282 - -
          Affymetrix Rat Clariom S Assay PSR0700020848.rn.1 TC0700002667.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10902290 - -
          Affymetrix Rat Gene 1.0 ST Array 10902282 - -
          Affymetrix Rat Gene 1.0 ST Array 10902280 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358360 - -
          RatRef-12 Expression BeadChip ILMN_51735 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline