Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01503968_g1
          See other DNAL4 GE Assays ›
          Gene Symbol:
          Dnal4
          Gene Name:
          dynein, axonemal, light chain 4
          Gene Aliases:
          Dnalc4
          Chromosome Location:
          Chr.7: 120987438 - 121000272 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01503968_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnal4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009666.1 NP_001009666.1 2 - 3 293 69
          GenBank mRNA BC087688.1 - 2 - 3 293 7311257 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnal4

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s)

          microtubule-based process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P106992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75370 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106992 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106992 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P106992 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208849 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P723060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74151 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905490 - -
          Affymetrix Rat Gene 1.0 ST Array 10905485 - -
          Affymetrix Rat Gene 1.0 ST Array 10905487 - -
          Affymetrix Rat Gene 1.0 ST Array 10905488 - -
          Affymetrix Rat Gene 1.0 ST Array 10905486 - -
          Affymetrix Rat Gene 1.0 ST Array 10905491 - -
          Affymetrix Rat Gene 1.0 ST Array 10905489 - -
          Affymetrix Rat Gene 1.1 ST Array 10905484 - -
          Rat Genome 230 2.0 Array 1388785_at - -
          Rat Genome 230 2.0 Array 1372514_PM_s_at - -
          Rat Genome 230 2.0 Array 1388785_PM_at - -
          Rat Genome 230 2.0 Array 1372514_s_at - -
          Affymetrix HT Rat Focus Array 1388785_at - -
          Affymetrix HT Rat Focus Array 1388785_PM_at - -
          Affymetrix HT Rat Focus Array 1372514_PM_s_at - -
          Affymetrix HT Rat Focus Array 1372514_s_at - -
          Affymetrix Rat Clariom S Assay PSR0700027689.rn.1 TC0700003511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027690.rn.1 TC0700003511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027688.rn.1 TC0700003511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027691.rn.1 TC0700003511.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027692.rn.1 TC0700003511.rn.1 RTA TC
          Rat Expression Set 230 1372514_s_at - -
          Rat Expression Set 230 1388785_PM_at - -
          Rat Expression Set 230 1372514_PM_s_at - -
          Rat Expression Set 230 1388785_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905484 - -
          Rat Genome U34 Set rc_AA892303_g_at - -
          Rat Genome U34 Set rc_AA892303_at - -
          Rat Genome U34 Set rc_AI011666_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21006671 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364188 - -
          RatRef-12 Expression BeadChip ILMN_51203 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline