Search
Search
| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053546.1 | NP_445998.1 | 6 - 7 | 1332 | 93 | |
| XM_006241609.2 | XP_006241671.1 | 6 - 7 | 1677 | 93 | ||
| XM_008765463.1 | XP_008763685.1 | 6 - 7 | 861 | 93 | ||
| XM_008765464.1 | XP_008763686.1 | 6 - 7 | 3325 | 93 | ||
| GenBank mRNA | AB080023.1 | - | 6 - 7 | 1332 | 93 | |
Set Membership: |
|