Search
Search
| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024876.1 | NP_001020047.1 | 2 - 3 | 234 | 67 | |
| XM_006241671.2 | XP_006241733.1 | 2 - 3 | 274 | 67 | ||
| XM_008765506.1 | XP_008763728.1 | 2 - 3 | 274 | 67 | ||
| GenBank mRNA | BC097477.1 | - | 2 - 3 | 234 | 7458086 | 67 |
Set Membership: |
|