Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505088_m1
          See other ENPP2 GE Assays ›
          Gene Symbol:
          Enpp2
          Gene Name:
          ectonucleotide pyrophosphatase/phosphodiesterase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 94479931 - 94563086 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Enpp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057104.2 NP_476445.2 21 - 22 2511 66
          GenBank mRNA BC081747.1 - 21 - 22 2471 7115236 66
          D28560.1 - 23 - 24 2580 66
          DQ131564.1 - 21 - 22 2511 66
          FQ233297.1 - 7 - 8 827 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Enpp2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell-matrix adhesion
          receptor-mediated endocytosis
          chemotaxis
          immune response
          positive regulation of cell proliferation
          phospholipid catabolic process
          regulation of cell migration
          phosphatidylcholine catabolic process
          estrous cycle
          regulation of angiogenesis
          positive regulation of oligodendrocyte differentiation
          positive regulation of focal adhesion assembly
          cell chemotaxis
          cellular response to cadmium ion
          cellular response to estradiol stimulus
          nucleic acid phosphodiester bond hydrolysis
          positive regulation of substrate adhesion-dependent cell spreading
          response to polycyclic arene
          nucleic acid binding
          phosphodiesterase I activity
          nucleotide diphosphatase activity
          lysophospholipase activity
          scavenger receptor activity
          calcium ion binding
          zinc ion binding
          polysaccharide binding
          metal ion binding
          alkylglycerophosphoethanolamine phosphodiesterase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P341075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52415 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12327 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61267 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P341075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P341075 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P341075 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P341075 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700304870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47797 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46540 - -
          Agilent Rat cDNA Microarray (G4105A) 700304870 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903754 - -
          Affymetrix Rat Gene 1.0 ST Array 10903752 - -
          Affymetrix Rat Gene 1.0 ST Array 10903741 - -
          Affymetrix Rat Gene 1.0 ST Array 10903742 - -
          Affymetrix Rat Gene 1.0 ST Array 10903760 - -
          Affymetrix Rat Gene 1.0 ST Array 10903747 - -
          Affymetrix Rat Gene 1.0 ST Array 10903740 - -
          Affymetrix Rat Gene 1.0 ST Array 10903762 - -
          Affymetrix Rat Gene 1.0 ST Array 10903743 - -
          Affymetrix Rat Gene 1.0 ST Array 10903748 - -
          Affymetrix Rat Gene 1.0 ST Array 10903738 - -
          Affymetrix Rat Gene 1.0 ST Array 10903749 - -
          Affymetrix Rat Gene 1.0 ST Array 10903746 - -
          Affymetrix Rat Gene 1.0 ST Array 10903755 - -
          Affymetrix Rat Gene 1.0 ST Array 10903761 - -
          Affymetrix Rat Gene 1.0 ST Array 10903759 - -
          Affymetrix Rat Gene 1.0 ST Array 10903744 - -
          Affymetrix Rat Gene 1.0 ST Array 10903737 - -
          Affymetrix Rat Gene 1.0 ST Array 10903739 - -
          Affymetrix Rat Gene 1.0 ST Array 10903758 - -
          Affymetrix Rat Gene 1.0 ST Array 10903757 - -
          Affymetrix Rat Gene 1.0 ST Array 10903756 - -
          Affymetrix Rat Gene 1.0 ST Array 10903750 - -
          Affymetrix Rat Gene 1.0 ST Array 10903753 - -
          Affymetrix Rat Gene 1.0 ST Array 10903745 - -
          Affymetrix Rat Gene 1.1 ST Array 10903736 - -
          Rat Genome 230 2.0 Array 1368536_PM_at - -
          Rat Genome 230 2.0 Array 1368536_at - -
          Affymetrix HT Rat Focus Array 1368536_PM_at - -
          Affymetrix HT Rat Focus Array 1368536_at - -
          Rat Expression Set 230 1368536_at - -
          Rat Expression Set 230 1368536_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903736 - -
          Rat Genome U34 Set D28560_g_at - -
          Rat Genome U34 Set D28560_at - -
          Rat Genome U34 Set D28560_at - -
          Rat Genome U34 Set D28560_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21378132 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline