Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505234_m1
          See other GGA1 GE Assays ›
          Gene Symbol:
          Gga1
          Gene Name:
          golgi associated, gamma adaptin ear containing, ARF binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 120102592 - 120119738 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505234_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gga1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011994.1 NP_001011994.1 4 - 5 526 71
          XM_006241973.2 XP_006242035.1 5 - 6 1888 71
          XM_008765679.1 XP_008763901.1 6 - 7 1644 71
          XM_008765680.1 XP_008763902.1 3 - 4 1142 71
          XM_008765681.1 XP_008763903.1 5 - 6 4350 71
          GenBank mRNA BC090031.1 - 4 - 5 526 7366311 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001430.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gga1

          Gene Ontology Categories:

          Function(s)

          intracellular protein transport
          vesicle-mediated transport
          positive regulation of protein catabolic process
          toxin transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P081451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081451 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P236806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20940 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897493 - -
          Affymetrix Rat Gene 1.0 ST Array 10897494 - -
          Affymetrix Rat Gene 1.0 ST Array 10897487 - -
          Affymetrix Rat Gene 1.0 ST Array 10897492 - -
          Affymetrix Rat Gene 1.0 ST Array 10897503 - -
          Affymetrix Rat Gene 1.0 ST Array 10897502 - -
          Affymetrix Rat Gene 1.0 ST Array 10897500 - -
          Affymetrix Rat Gene 1.0 ST Array 10897499 - -
          Affymetrix Rat Gene 1.0 ST Array 10897495 - -
          Affymetrix Rat Gene 1.0 ST Array 10897488 - -
          Affymetrix Rat Gene 1.0 ST Array 10897489 - -
          Affymetrix Rat Gene 1.0 ST Array 10897498 - -
          Affymetrix Rat Gene 1.0 ST Array 10897490 - -
          Affymetrix Rat Gene 1.0 ST Array 10897496 - -
          Affymetrix Rat Gene 1.0 ST Array 10897497 - -
          Affymetrix Rat Gene 1.0 ST Array 10897491 - -
          Affymetrix Rat Gene 1.0 ST Array 10897501 - -
          Affymetrix Rat Gene 1.1 ST Array 10897485 - -
          Rat Genome 230 2.0 Array 1374761_PM_at - -
          Rat Genome 230 2.0 Array 1374761_at - -
          Affymetrix HT Rat Focus Array 1374761_at - -
          Affymetrix HT Rat Focus Array 1374761_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700010593.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010597.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010599.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010600.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010587.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010589.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010602.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010591.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010592.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010594.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010598.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010588.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010596.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010601.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010585.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010595.rn.1 TC0700001430.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010603.rn.1 TC0700001430.rn.1 RTA TC
          Rat Expression Set 230 1374761_at - -
          Rat Expression Set 230 1374761_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897485 - -
          Rat Genome U34 Set rc_AI029572_at - -
          Rat Genome U34 Set rc_AI175216_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21316062 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376859 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375809 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline