Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505402_m1
          See other ATAD2 GE Assays ›
          Gene Symbol:
          Atad2
          Gene Name:
          ATPase family, AAA domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 97984754 - 98027379 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505402_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134879.1 NP_001128351.1 12 - 13 1619 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001154.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          positive regulation of transcription, DNA-templated
          ATP binding
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P385014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P385014 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P385014 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P385014 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P385014 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P284110 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P446375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903869 - -
          Affymetrix Rat Gene 1.0 ST Array 10903889 - -
          Affymetrix Rat Gene 1.0 ST Array 10903877 - -
          Affymetrix Rat Gene 1.0 ST Array 10903893 - -
          Affymetrix Rat Gene 1.0 ST Array 10903884 - -
          Affymetrix Rat Gene 1.0 ST Array 10903886 - -
          Affymetrix Rat Gene 1.0 ST Array 10903880 - -
          Affymetrix Rat Gene 1.0 ST Array 10903876 - -
          Affymetrix Rat Gene 1.0 ST Array 10903872 - -
          Affymetrix Rat Gene 1.0 ST Array 10903874 - -
          Affymetrix Rat Gene 1.0 ST Array 10903878 - -
          Affymetrix Rat Gene 1.0 ST Array 10903875 - -
          Affymetrix Rat Gene 1.0 ST Array 10903882 - -
          Affymetrix Rat Gene 1.0 ST Array 10903871 - -
          Affymetrix Rat Gene 1.0 ST Array 10903894 - -
          Affymetrix Rat Gene 1.0 ST Array 10903890 - -
          Affymetrix Rat Gene 1.0 ST Array 10903879 - -
          Affymetrix Rat Gene 1.0 ST Array 10903891 - -
          Affymetrix Rat Gene 1.0 ST Array 10903887 - -
          Affymetrix Rat Gene 1.0 ST Array 10903888 - -
          Affymetrix Rat Gene 1.0 ST Array 10903885 - -
          Affymetrix Rat Gene 1.0 ST Array 10903892 - -
          Affymetrix Rat Gene 1.0 ST Array 10903873 - -
          Affymetrix Rat Gene 1.0 ST Array 10903870 - -
          Affymetrix Rat Gene 1.0 ST Array 10903881 - -
          Affymetrix Rat Gene 1.1 ST Array 10903864 - -
          Rat Genome 230 2.0 Array 1376599_PM_at - -
          Rat Genome 230 2.0 Array 1391651_PM_at - -
          Rat Genome 230 2.0 Array 1391651_at - -
          Rat Genome 230 2.0 Array 1376599_at - -
          Affymetrix HT Rat Focus Array 1376599_at - -
          Affymetrix HT Rat Focus Array 1391651_PM_at - -
          Affymetrix HT Rat Focus Array 1376599_PM_at - -
          Affymetrix HT Rat Focus Array 1391651_at - -
          Affymetrix Rat Clariom S Assay PSR0700008425.rn.1 TC0700001154.rn.1 RTA TC
          Rat Expression Set 230 1376599_at - -
          Rat Expression Set 230 1376599_PM_at - -
          Rat Expression Set 230 1391651_at - -
          Rat Expression Set 230 1391651_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903864 - -
          Rat Genome U34 Set rc_AA801178_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22106362 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650533 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline