Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505497_m1
          See other SAMM50 GE Assays ›
          Gene Symbol:
          Samm50
          Gene Name:
          SAMM50 sorting and assembly machinery component
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 125058966 - 125082506 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505497_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Samm50
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004241.1 NP_001004241.1 14 - 15 1420 80
          GenBank mRNA BC079302.1 - 14 - 15 1420 7134547 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001563.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Samm50

          Gene Ontology Categories:

          Function(s)

          protein import into mitochondrial outer membrane

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P231399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108731 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231399 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231399 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P231399 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231399 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1056278 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898226 - -
          Affymetrix Rat Gene 1.0 ST Array 10898225 - -
          Affymetrix Rat Gene 1.0 ST Array 10898218 - -
          Affymetrix Rat Gene 1.0 ST Array 10898219 - -
          Affymetrix Rat Gene 1.0 ST Array 10898227 - -
          Affymetrix Rat Gene 1.0 ST Array 10898214 - -
          Affymetrix Rat Gene 1.0 ST Array 10898216 - -
          Affymetrix Rat Gene 1.0 ST Array 10898213 - -
          Affymetrix Rat Gene 1.0 ST Array 10898215 - -
          Affymetrix Rat Gene 1.0 ST Array 10898222 - -
          Affymetrix Rat Gene 1.0 ST Array 10898223 - -
          Affymetrix Rat Gene 1.0 ST Array 10898217 - -
          Affymetrix Rat Gene 1.0 ST Array 10898224 - -
          Affymetrix Rat Gene 1.0 ST Array 10898228 - -
          Affymetrix Rat Gene 1.0 ST Array 10898220 - -
          Affymetrix Rat Gene 1.0 ST Array 10898221 - -
          Affymetrix Rat Gene 1.1 ST Array 10898212 - -
          Rat Genome 230 2.0 Array 1388380_at - -
          Rat Genome 230 2.0 Array 1388380_PM_at - -
          Affymetrix HT Rat Focus Array 1388380_at - -
          Affymetrix HT Rat Focus Array 1388380_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700011888.rn.1 TC0700001563.rn.1 RTA TC
          Rat Expression Set 230 1388380_at - -
          Rat Expression Set 230 1388380_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898212 - -
          Rat Genome U34 Set rc_AI232741_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356478 - -
          RatRef-12 Expression BeadChip ILMN_49990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline