Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505519_m1
          See other ATP6V1C1 GE Assays ›
          Gene Symbol:
          Atp6v1c1
          Gene Name:
          ATPase, H+ transporting, lysosomal V1 subunit C1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 77678864 - 77716658 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505519_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6v1c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011992.1 NP_001011992.1 12 - 13 1151 86
          XM_006241602.2 XP_006241664.1 11 - 12 1156 86
          GenBank mRNA BC089961.1 - 12 - 13 1151 7314200 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001011.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp6v1c1

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          ATP hydrolysis coupled proton transport
          hydrogen ion transmembrane transport
          molecular_function
          hydrogen-exporting ATPase activity, phosphorylative mechanism

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P022263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110673 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167322 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022263 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P022263 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P022263 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54777 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71919 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37150 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896245 - -
          Affymetrix Rat Gene 1.0 ST Array 10896242 - -
          Affymetrix Rat Gene 1.0 ST Array 10896243 - -
          Affymetrix Rat Gene 1.0 ST Array 10896237 - -
          Affymetrix Rat Gene 1.0 ST Array 10896240 - -
          Affymetrix Rat Gene 1.0 ST Array 10896249 - -
          Affymetrix Rat Gene 1.0 ST Array 10896235 - -
          Affymetrix Rat Gene 1.0 ST Array 10896241 - -
          Affymetrix Rat Gene 1.0 ST Array 10896238 - -
          Affymetrix Rat Gene 1.0 ST Array 10896248 - -
          Affymetrix Rat Gene 1.0 ST Array 10896244 - -
          Affymetrix Rat Gene 1.0 ST Array 10896247 - -
          Affymetrix Rat Gene 1.0 ST Array 10896246 - -
          Affymetrix Rat Gene 1.0 ST Array 10896236 - -
          Affymetrix Rat Gene 1.0 ST Array 10896239 - -
          Affymetrix Rat Gene 1.1 ST Array 10896234 - -
          Rat Genome 230 2.0 Array 1374396_PM_at - -
          Rat Genome 230 2.0 Array 1374396_at - -
          Affymetrix HT Rat Focus Array 1374396_PM_at - -
          Affymetrix HT Rat Focus Array 1374396_at - -
          Affymetrix Rat Clariom S Assay PSR0700007596.rn.1 TC0700001011.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700007595.rn.1 TC0700001011.rn.1 RTA TC
          Rat Expression Set 230 1374396_PM_at - -
          Rat Expression Set 230 1374396_at - -
          Affymetrix Rat Gene 1.0 ST Array 10896234 - -
          Rat Genome U34 Set rc_AA799824_at - -
          Rat Genome U34 Set rc_AA819556_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21758866 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375808 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376957 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline