Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505834_m1
          See other RSPO2 GE Assays ›
          Gene Symbol:
          Rspo2
          Gene Name:
          R-spondin 2
          Gene Aliases:
          RGD1562331
          Chromosome Location:
          Chr.7: 81919911 - 82059788 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505834_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rspo2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130575.1 NP_001124047.1 3 - 4 1425 74
          XM_008765454.1 XP_008763676.1 3 - 4 1575 74
          XM_008765455.1 XP_008763677.1 3 - 4 874 74
          GenBank mRNA AB232173.1 - 3 - 4 1577 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003070.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          biological_process
          bone mineralization
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          negative regulation of odontogenesis of dentin-containing tooth
          lung growth
          epithelial tube branching involved in lung morphogenesis
          trachea cartilage morphogenesis
          dopaminergic neuron differentiation
          positive regulation of canonical Wnt signaling pathway
          G-protein coupled receptor binding
          molecular_function
          receptor binding
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082924 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082919 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P610634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903541 - -
          Affymetrix Rat Gene 1.0 ST Array 10903542 - -
          Affymetrix Rat Gene 1.0 ST Array 10903543 - -
          Affymetrix Rat Gene 1.0 ST Array 10903544 - -
          Affymetrix Rat Gene 1.0 ST Array 10903540 - -
          Affymetrix Rat Gene 1.0 ST Array 10903539 - -
          Affymetrix Rat Gene 1.1 ST Array 10903538 - -
          Affymetrix Rat Clariom S Assay PSR0700023671.rn.1 TC0700003070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700023672.rn.1 TC0700003070.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10903538 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368734 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline