Search
Search
| Gene Symbol: Grhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134527.1 | NP_001127999.1 | 4 - 5 | 818 | 66 | |
| XM_006241552.2 | XP_006241614.1 | 4 - 5 | 1646 | 66 | ||
| GenBank mRNA | BC168688.1 | - | 4 - 5 | 818 | 7932577 | 66 |
Set Membership: |
|