Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01506215_m1
          See other PRR5 GE Assays ›
          Gene Symbol:
          Prr5
          Gene Name:
          proline rich 5 (renal)
          Gene Aliases:
          Protor1
          Chromosome Location:
          Chr.7: 125569868 - 125612287 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01506215_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012121.1 NP_001012121.1 7 - 8 867 87
          XM_008765722.1 XP_008763944.1 8 - 9 1963 87
          XM_008765723.1 XP_008763945.1 7 - 8 723 87
          XM_008765724.1 XP_008763946.1 7 - 8 1548 87
          XM_008765725.1 XP_008763947.1 8 - 9 700 87
          XM_008765726.1 XP_008763948.1 8 - 9 695 87
          GenBank mRNA BC090007.1 - 7 - 8 867 7323849 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001568.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr5

          Gene Ontology Categories:

          Function(s)

          positive regulation of protein phosphorylation
          positive regulation of phosphatidylinositol 3-kinase signaling

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P165451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127563 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135349 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165451 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P165451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P331097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69781 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898265 - -
          Affymetrix Rat Gene 1.0 ST Array 10898266 - -
          Affymetrix Rat Gene 1.0 ST Array 10898263 - -
          Affymetrix Rat Gene 1.0 ST Array 10898262 - -
          Affymetrix Rat Gene 1.0 ST Array 10898267 - -
          Affymetrix Rat Gene 1.0 ST Array 10898261 - -
          Affymetrix Rat Gene 1.0 ST Array 10898264 - -
          Affymetrix Rat Gene 1.0 ST Array 10898260 - -
          Affymetrix Rat Gene 1.1 ST Array 10898259 - -
          Rat Genome 230 2.0 Array 1392563_at - -
          Rat Genome 230 2.0 Array 1392563_PM_at - -
          Affymetrix HT Rat Focus Array 1392563_PM_at - -
          Affymetrix HT Rat Focus Array 1392563_at - -
          Affymetrix Rat Clariom S Assay PSR0700011963.rn.1 TC0700001568.rn.1 RTA TC
          Rat Expression Set 230 1392563_PM_at - -
          Rat Expression Set 230 1392563_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898259 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376406 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline