Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01506370_m1
          See other UQCRC2 GE Assays ›
          Gene Symbol:
          Uqcrc2
          Gene Name:
          ubiquinol cytochrome c reductase core protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 190555177 - 190585741 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01506370_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Uqcrc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006970.1 NP_001006971.1 13 - 14 1310 98
          GenBank mRNA BC083610.1 - 13 - 14 1310 7133503 98
          FQ219943.1 - 5 - 6 591 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002502.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Uqcrc2

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          oxidation-reduction process
          metalloendopeptidase activity
          protein complex binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P379647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379647 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P379647 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P379647 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P379647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94864 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92784 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94010 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710438 - -
          Affymetrix Rat Gene 1.0 ST Array 10710434 - -
          Affymetrix Rat Gene 1.0 ST Array 10710431 - -
          Affymetrix Rat Gene 1.0 ST Array 10710440 - -
          Affymetrix Rat Gene 1.0 ST Array 10710429 - -
          Affymetrix Rat Gene 1.0 ST Array 10710441 - -
          Affymetrix Rat Gene 1.0 ST Array 10710432 - -
          Affymetrix Rat Gene 1.0 ST Array 10710430 - -
          Affymetrix Rat Gene 1.0 ST Array 10710439 - -
          Affymetrix Rat Gene 1.0 ST Array 10710435 - -
          Affymetrix Rat Gene 1.0 ST Array 10710428 - -
          Affymetrix Rat Gene 1.0 ST Array 10710433 - -
          Affymetrix Rat Gene 1.0 ST Array 10725377 - -
          Affymetrix Rat Gene 1.0 ST Array 10710436 - -
          Affymetrix Rat Gene 1.0 ST Array 10710437 - -
          Affymetrix Rat Gene 1.1 ST Array 10725376 - -
          Affymetrix Rat Gene 1.1 ST Array 10710427 - -
          Rat Genome 230 2.0 Array 1382280_at - -
          Rat Genome 230 2.0 Array 1373362_at - -
          Rat Genome 230 2.0 Array 1382280_PM_at - -
          Rat Genome 230 2.0 Array 1373362_PM_at - -
          Affymetrix HT Rat Focus Array 1373362_PM_at - -
          Affymetrix HT Rat Focus Array 1373362_at - -
          Affymetrix HT Rat Focus Array 1382280_PM_at - -
          Affymetrix HT Rat Focus Array 1382280_at - -
          Affymetrix Rat Clariom S Assay PSR0100019211.rn.1 TC0100002502.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100019212.rn.1 TC0100002502.rn.1 RTA TC
          Rat Expression Set 230 1373362_PM_at - -
          Rat Expression Set 230 1373362_at - -
          Rat Expression Set 230 1382280_PM_at - -
          Rat Expression Set 230 1382280_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710427 - -
          Rat Genome U34 Set rc_AI235358_at - -
          Rat Genome U34 Set rc_AI233114_at - -
          Rat Genome U34 Set rc_AI235358_at - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_63688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline