Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01506504_m1
          See other LYRM1 GE Assays ›
          Gene Symbol:
          Lyrm1
          Gene Name:
          LYR motif containing 1
          Gene Aliases:
          RGD1563498
          Chromosome Location:
          Chr.1: 189663200 - 189685842 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01506504_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyrm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001276487.1 NP_001263416.1 2 - 3 261 152
          XM_006230132.2 XP_006230194.1 1 - 2 201 152
          XM_006230133.1 XP_006230195.1 1 - 2 197 152
          XM_006230134.2 XP_006230196.2 2 - 3 2504 152
          GenBank mRNA BC166909.1 - 2 - 3 258 6889767 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002489.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458511 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104870 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104870 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P123785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61368 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710335 - -
          Affymetrix Rat Gene 1.0 ST Array 10710334 - -
          Affymetrix Rat Gene 1.0 ST Array 10710336 - -
          Affymetrix Rat Gene 1.0 ST Array 10710337 - -
          Affymetrix Rat Gene 1.1 ST Array 10710333 - -
          Rat Genome 230 2.0 Array 1389300_at - -
          Rat Genome 230 2.0 Array 1389300_PM_at - -
          Affymetrix HT Rat Focus Array 1389300_PM_at - -
          Affymetrix HT Rat Focus Array 1389300_at - -
          Affymetrix Rat Clariom S Assay PSR0100018976.rn.1 TC0100002489.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018979.rn.1 TC0100002489.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018978.rn.1 TC0100002489.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018975.rn.1 TC0100002489.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018977.rn.1 TC0100002489.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100018973.rn.1 TC0100002489.rn.1 RTA TC
          Rat Expression Set 230 1389300_PM_at - -
          Rat Expression Set 230 1389300_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710333 - -
          Rat Genome U34 Set rc_AI009014_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20902780 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371750 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline