Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01506548_m1
          See other ACSM3 GE Assays ›
          Gene Symbol:
          Acsm3
          Gene Name:
          acyl-CoA synthetase medium-chain family member 3
          Gene Aliases:
          Sa, Sah
          Chromosome Location:
          Chr.1: 189514503 - 189541233 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01506548_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033231.1 NP_150234.1 13 - 14 1826 138
          XM_006230105.2 XP_006230167.1 13 - 14 1848 138
          XM_006230106.2 XP_006230168.1 13 - 14 1809 138
          GenBank mRNA BC090325.1 - 13 - 14 1826 7128525 138
          FQ209573.1 - 5 - 6 486 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002484.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acsm3

          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid biosynthetic process
          biological_process
          molecular_function
          ATP binding
          fatty acid ligase activity
          metal ion binding
          butyrate-CoA ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15937 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15937 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15937 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15937 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700887302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87961 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58853 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87640 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46745 - -
          Agilent Rat cDNA Microarray (G4105A) 700887302 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710305 - -
          Affymetrix Rat Gene 1.0 ST Array 10710301 - -
          Affymetrix Rat Gene 1.0 ST Array 10710296 - -
          Affymetrix Rat Gene 1.0 ST Array 10710297 - -
          Affymetrix Rat Gene 1.0 ST Array 10710300 - -
          Affymetrix Rat Gene 1.0 ST Array 10710310 - -
          Affymetrix Rat Gene 1.0 ST Array 10710308 - -
          Affymetrix Rat Gene 1.0 ST Array 10710299 - -
          Affymetrix Rat Gene 1.0 ST Array 10710304 - -
          Affymetrix Rat Gene 1.0 ST Array 10710303 - -
          Affymetrix Rat Gene 1.0 ST Array 10710312 - -
          Affymetrix Rat Gene 1.0 ST Array 10710311 - -
          Affymetrix Rat Gene 1.0 ST Array 10710298 - -
          Affymetrix Rat Gene 1.0 ST Array 10710302 - -
          Affymetrix Rat Gene 1.0 ST Array 10710309 - -
          Affymetrix Rat Gene 1.0 ST Array 10710306 - -
          Affymetrix Rat Gene 1.1 ST Array 10710295 - -
          Rat Genome 230 2.0 Array 1383303_at - -
          Rat Genome 230 2.0 Array 1383303_PM_at - -
          Affymetrix HT Rat Focus Array 1383303_PM_at - -
          Affymetrix HT Rat Focus Array 1383303_at - -
          Affymetrix Rat Clariom S Assay PSR0100018889.rn.1 TC0100002484.rn.1 RTA TC
          Rat Expression Set 230 1383303_at - -
          Rat Expression Set 230 1383303_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710295 - -
          Rat Genome U34 Set AF027188UTR#1_at - -
          Rat Genome U34 Set S62516_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21162411 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351101 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline