Search
Search

Plekha1| Gene Symbol: Plekha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079894.1 | NP_001073363.1 | 7 - 8 | 734 | 72 | |
| XM_006230409.2 | XP_006230471.1 | 7 - 8 | 812 | 72 | ||
| XM_006230412.2 | XP_006230474.1 | 7 - 8 | 813 | 72 | ||
| XM_006230413.2 | XP_006230475.1 | 7 - 8 | 812 | 72 | ||
| XM_006230415.2 | XP_006230477.1 | 7 - 8 | 812 | 72 | ||
| XM_008759907.1 | XP_008758129.1 | 7 - 8 | 887 | 72 | ||
| XM_008759908.1 | XP_008758130.1 | 7 - 8 | 1208 | 72 | ||
| XM_008759909.1 | XP_008758131.1 | 7 - 8 | 781 | 72 | ||
| XM_008759910.1 | XP_008758132.1 | 4 - 5 | 451 | 72 | ||
| GenBank mRNA | BC128752.1 | - | 7 - 8 | 734 | 7100072 | 72 |
Set Membership: |
|