Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01506787_m1
          See other DGAT2 GE Assays ›
          Gene Symbol:
          Dgat2
          Gene Name:
          diacylglycerol O-acyltransferase 2
          Gene Aliases:
          ARAT
          Chromosome Location:
          Chr.1: 164113459 - 164143818 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01506787_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dgat2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012345.1 NP_001012345.1 7 - 8 1187 58
          XM_006229733.2 XP_006229795.1 6 - 7 1227 58
          XM_008759689.1 XP_008757911.1 7 - 8 938 58
          GenBank mRNA BC089846.1 - 7 - 8 1187 7372077 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006110.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dgat2

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          triglyceride biosynthetic process
          lipid storage
          low-density lipoprotein particle clearance
          long-chain fatty-acyl-CoA metabolic process
          cellular triglyceride homeostasis
          retinol metabolic process
          cholesterol homeostasis
          diacylglycerol metabolic process
          fatty acid homeostasis
          fat pad development
          cellular response to oleic acid
          regulation of plasma lipoprotein particle levels
          2-acylglycerol O-acyltransferase activity
          diacylglycerol O-acyltransferase activity
          protein homodimerization activity
          retinol O-fatty-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P733209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P733209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P733209 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P733209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15649 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P733209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P180012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723874 - -
          Affymetrix Rat Gene 1.0 ST Array 10723870 - -
          Affymetrix Rat Gene 1.0 ST Array 10723868 - -
          Affymetrix Rat Gene 1.0 ST Array 10723867 - -
          Affymetrix Rat Gene 1.0 ST Array 10723871 - -
          Affymetrix Rat Gene 1.0 ST Array 10723873 - -
          Affymetrix Rat Gene 1.0 ST Array 10723872 - -
          Affymetrix Rat Gene 1.0 ST Array 10723869 - -
          Affymetrix Rat Gene 1.1 ST Array 10723866 - -
          Rat Genome 230 2.0 Array 1391045_at - -
          Rat Genome 230 2.0 Array 1371615_PM_at - -
          Rat Genome 230 2.0 Array 1371615_at - -
          Rat Genome 230 2.0 Array 1391045_PM_at - -
          Affymetrix HT Rat Focus Array 1371615_at - -
          Affymetrix HT Rat Focus Array 1391045_at - -
          Affymetrix HT Rat Focus Array 1391045_PM_at - -
          Affymetrix HT Rat Focus Array 1371615_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100048475.rn.1 TC0100006110.rn.1 RTA TC
          Rat Expression Set 230 1371615_at - -
          Rat Expression Set 230 1371615_PM_at - -
          Rat Expression Set 230 1391045_PM_at - -
          Rat Expression Set 230 1391045_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723866 - -
          Rat Genome U34 Set rc_AI012505_at - -
          Rat Genome U34 Set rc_AI045040_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21388648 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376726 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline