Search
Search
| Gene Symbol: Numa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100691.1 | NP_001094161.1 | 18 - 19 | 5559 | 60 | |
| XM_008759723.1 | XP_008757945.1 | 19 - 20 | 5436 | 60 | ||
| GenBank mRNA | BC098753.1 | - | 7 - 8 | 1039 | 60 | |
Set Membership: |
|