Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000141.1 | NP_001000141.1 | 1 | 674 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |