Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01507619_g1
          See other CD19 GE Assays ›
          Gene Symbol:
          Cd19
          Gene Name:
          CD19 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 197815422 - 197822125 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01507619_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006230316.2 XP_006230378.1 13 - 14 1877 66
          GenBank mRNA BC090026.1 - 1 - 2 104 66

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          B cell receptor signaling pathway
          positive regulation of release of sequestered calcium ion into cytosol

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72684 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006898 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006898 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006898 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P156697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102285 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725695 - -
          Affymetrix Rat Gene 1.0 ST Array 10725692 - -
          Affymetrix Rat Gene 1.0 ST Array 10725697 - -
          Affymetrix Rat Gene 1.0 ST Array 10725689 - -
          Affymetrix Rat Gene 1.0 ST Array 10725686 - -
          Affymetrix Rat Gene 1.0 ST Array 10725700 - -
          Affymetrix Rat Gene 1.0 ST Array 10725699 - -
          Affymetrix Rat Gene 1.0 ST Array 10725690 - -
          Affymetrix Rat Gene 1.0 ST Array 10725687 - -
          Affymetrix Rat Gene 1.0 ST Array 10725693 - -
          Affymetrix Rat Gene 1.0 ST Array 10725691 - -
          Affymetrix Rat Gene 1.0 ST Array 10725696 - -
          Affymetrix Rat Gene 1.0 ST Array 10725698 - -
          Affymetrix Rat Gene 1.0 ST Array 10725694 - -
          Affymetrix Rat Gene 1.0 ST Array 10725688 - -
          Affymetrix Rat Gene 1.0 ST Array 10725685 - -
          Affymetrix Rat Gene 1.1 ST Array 10725684 - -
          Affymetrix Rat Gene 1.0 ST Array 10725684 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21569194 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367276 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline