Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01507631_m1
          See other RGS10 GE Assays ›
          Gene Symbol:
          Rgs10
          Gene Name:
          regulator of G-protein signaling 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 199782076 - 199823439 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01507631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgs10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019337.1 NP_062210.1 2 - 3 181 68
          XM_006230325.2 XP_006230387.1 2 - 3 222 68
          XM_006230326.2 XP_006230388.1 2 - 3 234 68
          GenBank mRNA CD372843.1 - 2 - 3 181 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006739.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          regulation of G-protein coupled receptor protein signaling pathway
          termination of G-protein coupled receptor signaling pathway
          positive regulation of GTPase activity
          molecular_function
          GTPase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130505 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095890 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095890 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342047 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700499481 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68990 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75331 - -
          Agilent Rat cDNA Microarray (G4105A) 700499481 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726139 - -
          Affymetrix Rat Gene 1.0 ST Array 10726137 - -
          Affymetrix Rat Gene 1.0 ST Array 10726136 - -
          Affymetrix Rat Gene 1.0 ST Array 10726135 - -
          Affymetrix Rat Gene 1.0 ST Array 10726138 - -
          Affymetrix Rat Gene 1.1 ST Array 10726134 - -
          Rat Genome 230 2.0 Array 1390707_at - -
          Rat Genome 230 2.0 Array 1390707_PM_at - -
          Affymetrix HT Rat Focus Array 1390707_PM_at - -
          Affymetrix HT Rat Focus Array 1390707_at - -
          Affymetrix Rat Clariom S Assay PSR0100053195.rn.1 TC0100006739.rn.1 RTA TC
          Rat Expression Set 230 1390707_at - -
          Rat Expression Set 230 1390707_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726134 - -
          Rat Genome U34 Set rc_AA801173_at - -
          Rat Genome U34 Set rc_AI105257_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21234195 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline