Search
Search
| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013900.1 | NP_001013922.1 | 1 - 2 | 205 | 96 | |
| XM_006230243.1 | XP_006230305.1 | 1 - 2 | 134 | 96 | ||
| XM_006230244.1 | XP_006230306.1 | 1 - 2 | 131 | 96 | ||
| XM_006230245.1 | XP_006230307.1 | 1 - 2 | 131 | 96 | ||
| GenBank mRNA | BC079045.1 | - | 1 - 2 | 205 | 7115071 | 96 |
Set Membership: |
|