Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01508038_m1
          See other TMEM135 GE Assays ›
          Gene Symbol:
          Tmem135
          Gene Name:
          transmembrane protein 135
          Gene Aliases:
          PMP52, RGD1309948
          Chromosome Location:
          Chr.1: 152886979 - 153096180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01508038_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem135
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013896.1 NP_001013918.1 14 - 15 1407 97
          GenBank mRNA BC085115.1 - 14 - 15 1407 7106476 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006015.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem135

          Gene Ontology Categories:

          Function(s)

          peroxisome organization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177553 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177740 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99111 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169175 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164925 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158576 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86088 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 821 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073683 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073683 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299699 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54279 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42094 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97363 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61268 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25257 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86618 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723566 - -
          Affymetrix Rat Gene 1.0 ST Array 10723574 - -
          Affymetrix Rat Gene 1.0 ST Array 10723564 - -
          Affymetrix Rat Gene 1.0 ST Array 10723568 - -
          Affymetrix Rat Gene 1.0 ST Array 10723573 - -
          Affymetrix Rat Gene 1.0 ST Array 10723575 - -
          Affymetrix Rat Gene 1.0 ST Array 10723567 - -
          Affymetrix Rat Gene 1.0 ST Array 10723569 - -
          Affymetrix Rat Gene 1.0 ST Array 10723571 - -
          Affymetrix Rat Gene 1.0 ST Array 10723561 - -
          Affymetrix Rat Gene 1.0 ST Array 10723562 - -
          Affymetrix Rat Gene 1.0 ST Array 10723565 - -
          Affymetrix Rat Gene 1.0 ST Array 10723572 - -
          Affymetrix Rat Gene 1.0 ST Array 10723570 - -
          Affymetrix Rat Gene 1.0 ST Array 10723563 - -
          Affymetrix Rat Gene 1.1 ST Array 10723560 - -
          Rat Genome 230 2.0 Array 1392466_at - -
          Rat Genome 230 2.0 Array 1392466_PM_at - -
          Affymetrix HT Rat Focus Array 1392466_PM_at - -
          Affymetrix HT Rat Focus Array 1392466_at - -
          Affymetrix Rat Clariom S Assay PSR0100047793.rn.1 TC0100006015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047795.rn.1 TC0100006015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047796.rn.1 TC0100006015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047794.rn.1 TC0100006015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047797.rn.1 TC0100006015.rn.1 RTA TC
          Rat Expression Set 230 1392466_at - -
          Rat Expression Set 230 1392466_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723560 - -
          Rat Genome U34 Set rc_AI175635_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20921285 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649782 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline