Search
Search
| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079885.1 | NP_001073354.1 | 16 - 17 | 1625 | 65 | |
| NM_001198653.1 | NP_001185582.1 | 16 - 17 | 1625 | 65 | ||
| XM_006230159.2 | XP_006230221.1 | 16 - 17 | 3029 | 65 | ||
| GenBank mRNA | BC128762.1 | - | 16 - 17 | 1625 | 7113062 | 65 |
Set Membership: |
|