Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01508441_m1
          See other COG7 GE Assays ›
          Gene Symbol:
          Cog7
          Gene Name:
          component of oligomeric golgi complex 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 191889332 - 191948977 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01508441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cog7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033889.1 NP_001029061.1 15 - 16 2104 67
          GenBank mRNA BC101931.1 - 15 - 16 2104 7938773 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006555.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cog7

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          intracellular protein transport
          retrograde vesicle-mediated transport, Golgi to ER
          biological_process
          protein localization to organelle
          protein localization to Golgi apparatus
          protein stabilization
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553286 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140612 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001892 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001892 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001892 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P725303 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67636 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27050 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97596 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38860 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42237 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725442 - -
          Affymetrix Rat Gene 1.0 ST Array 10725439 - -
          Affymetrix Rat Gene 1.0 ST Array 10725432 - -
          Affymetrix Rat Gene 1.0 ST Array 10725434 - -
          Affymetrix Rat Gene 1.0 ST Array 10725436 - -
          Affymetrix Rat Gene 1.0 ST Array 10725437 - -
          Affymetrix Rat Gene 1.0 ST Array 10725430 - -
          Affymetrix Rat Gene 1.0 ST Array 10725440 - -
          Affymetrix Rat Gene 1.0 ST Array 10725441 - -
          Affymetrix Rat Gene 1.0 ST Array 10725443 - -
          Affymetrix Rat Gene 1.0 ST Array 10725431 - -
          Affymetrix Rat Gene 1.0 ST Array 10725433 - -
          Affymetrix Rat Gene 1.0 ST Array 10725444 - -
          Affymetrix Rat Gene 1.0 ST Array 10725438 - -
          Affymetrix Rat Gene 1.0 ST Array 10725428 - -
          Affymetrix Rat Gene 1.0 ST Array 10725429 - -
          Affymetrix Rat Gene 1.0 ST Array 10725435 - -
          Affymetrix Rat Gene 1.1 ST Array 10725427 - -
          Rat Genome 230 2.0 Array 1379876_at - -
          Rat Genome 230 2.0 Array 1379876_PM_at - -
          Rat Genome 230 2.0 Array 1393490_at - -
          Rat Genome 230 2.0 Array 1393490_PM_at - -
          Affymetrix HT Rat Focus Array 1379876_PM_at - -
          Affymetrix HT Rat Focus Array 1393490_PM_at - -
          Affymetrix HT Rat Focus Array 1379876_at - -
          Affymetrix HT Rat Focus Array 1393490_at - -
          Affymetrix Rat Clariom S Assay PSR0100051433.rn.1 TC0100006555.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051432.rn.1 TC0100006555.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051434.rn.1 TC0100006555.rn.1 RTA TC
          Rat Expression Set 230 1379876_at - -
          Rat Expression Set 230 1393490_at - -
          Rat Expression Set 230 1379876_PM_at - -
          Rat Expression Set 230 1393490_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725427 - -
          Rat Genome U34 Set rc_AI029775_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21788581 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368076 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline