Search
Search

Tmem41b| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012358.1 | NP_001012358.1 | 2 - 3 | 361 | 126 | |
| XM_008759766.1 | XP_008757988.1 | 2 - 3 | 482 | 126 | ||
| GenBank mRNA | BC089866.1 | - | 2 - 3 | 361 | 7321590 | 126 |
| FQ228941.1 | - | 2 - 3 | 422 | 126 | ||
Set Membership: |
|