Search
Search
| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007750.1 | NP_001007751.1 | 5 - 6 | 883 | 61 | |
| XM_006241228.2 | XP_006241290.1 | 5 - 6 | 878 | 61 | ||
| XM_006241229.2 | XP_006241291.1 | 5 - 6 | 878 | 61 | ||
| XM_006241230.2 | XP_006241292.1 | 5 - 6 | 878 | 61 | ||
| XM_006241231.2 | XP_006241293.1 | 5 - 6 | 878 | 61 | ||
| GenBank mRNA | BC082074.1 | - | 5 - 6 | 883 | 7133149 | 61 |
Set Membership: |
|