Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01509997_g1
          See other RRP8 GE Assays ›
          Gene Symbol:
          Rrp8
          Gene Name:
          ribosomal RNA processing 8, methyltransferase, homolog (yeast)
          Gene Aliases:
          RGD1308302
          Chromosome Location:
          Chr.1: 170574670 - 170578693 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01509997_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rrp8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008346.1 NP_001008347.1 6 - 7 1318 83
          GenBank mRNA BC085119.1 - 6 - 7 1318 7108537 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006298.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rrp8

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing at rDNA
          transcription, DNA-templated
          rRNA processing
          chromatin modification
          methylation
          cellular response to glucose starvation
          regulation of transcription by glucose
          positive regulation of cell cycle arrest
          intrinsic apoptotic signaling pathway by p53 class mediator
          methyltransferase activity
          methylated histone binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97925 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069819 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069819 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069819 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337579 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724523 - -
          Affymetrix Rat Gene 1.0 ST Array 10724520 - -
          Affymetrix Rat Gene 1.0 ST Array 10724519 - -
          Affymetrix Rat Gene 1.0 ST Array 10724522 - -
          Affymetrix Rat Gene 1.0 ST Array 10724518 - -
          Affymetrix Rat Gene 1.0 ST Array 10724517 - -
          Affymetrix Rat Gene 1.0 ST Array 10724521 - -
          Affymetrix Rat Gene 1.1 ST Array 10724516 - -
          Rat Genome 230 2.0 Array 1376681_at - -
          Rat Genome 230 2.0 Array 1376681_PM_at - -
          Affymetrix HT Rat Focus Array 1376681_PM_at - -
          Affymetrix HT Rat Focus Array 1376681_at - -
          Affymetrix Rat Clariom S Assay PSR0100049700.rn.1 TC0100006298.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049701.rn.1 TC0100006298.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049703.rn.1 TC0100006298.rn.1 RTA TC
          Rat Expression Set 230 1376681_PM_at - -
          Rat Expression Set 230 1376681_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724516 - -
          Rat Genome U34 Set rc_AI177038_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349140 - -
          RatRef-12 Expression BeadChip ILMN_59493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline