Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100582.1 | NP_001094052.1 | 9 - 10 | 1134 | 54 | |
XM_006229953.2 | XP_006230015.1 | 7 - 8 | 1044 | 54 | ||
XM_006229955.2 | XP_006230017.1 | 5 - 6 | 1438 | 54 | ||
XM_008759733.1 | XP_008757955.1 | 9 - 10 | 1106 | 54 | ||
XM_008759734.1 | XP_008757956.1 | 10 - 11 | 1464 | 54 | ||
XM_008759735.1 | XP_008757957.1 | 8 - 9 | 1227 | 54 | ||
XM_008759736.1 | XP_008757958.1 | 6 - 7 | 1464 | 54 | ||
GenBank mRNA | AY754026.1 | - | 4 - 5 | 352 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |