Search
Search

Tmem159| Gene Symbol: Tmem159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194354.2 | NP_919335.2 | 3 - 4 | 266 | 88 | |
| XM_006230138.2 | XP_006230200.1 | 2 - 3 | 359 | 88 | ||
| XM_006230139.2 | XP_006230201.1 | 3 - 4 | 404 | 88 | ||
| XM_008759784.1 | XP_008758006.1 | 2 - 3 | 1197 | 88 | ||
| XM_008759785.1 | XP_008758007.1 | 2 - 3 | 1192 | 88 | ||
| GenBank mRNA | AY368496.1 | - | 1 - 2 | 100 | 88 | |
| BC076384.1 | - | 3 - 4 | 266 | 7107584 | 88 | |
Set Membership: |
|