Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01511015_m1
          See other LMO1 GE Assays ›
          Gene Symbol:
          Lmo1
          Gene Name:
          LIM domain only 1
          Gene Aliases:
          Dat1, Lmo3
          Chromosome Location:
          Chr.1: 173728183 - 173764288 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01511015_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lmo1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139112.2 NP_620812.2 2 - 3 363 76
          XM_006229968.1 XP_006230030.1 3 - 4 665 76
          GenBank mRNA BF420017.1 - 2 - 3 363 76
          BF567497.1 - 1 - 2 175 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006371.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lmo1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159088 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033845 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033845 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033845 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724712 - -
          Affymetrix Rat Gene 1.0 ST Array 10724711 - -
          Affymetrix Rat Gene 1.0 ST Array 10724709 - -
          Affymetrix Rat Gene 1.0 ST Array 10724710 - -
          Affymetrix Rat Gene 1.1 ST Array 10724708 - -
          Affymetrix Rat Clariom S Assay PSR0100049961.rn.1 TC0100006371.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10724708 - -
          Rat Genome U34 Set rc_AI236355_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22239669 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376311 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0