Search
Search
| Gene Symbol: Trim72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077675.1 | NP_001071143.1 | 2 - 3 | 481 | 79 | |
| GenBank mRNA | BC127539.1 | - | 2 - 3 | 481 | 7192024 | 79 |
| FQ224850.1 | - | 2 - 3 | 522 | 79 | ||
Set Membership: |
|