Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01511457_m1
          See other CDR2 GE Assays ›
          Gene Symbol:
          Cdr2
          Gene Name:
          cerebellar degeneration-related protein 2
          Gene Aliases:
          RGD1310578
          Chromosome Location:
          Chr.1: 190889698 - 190914606 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01511457_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025682.1 NP_001020853.1 2 - 3 264 91
          GenBank mRNA BC098028.1 - 2 - 3 264 7456103 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006538.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdr2

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158652 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8252 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018071 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018071 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018071 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32999 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4554 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725388 - -
          Affymetrix Rat Gene 1.0 ST Array 10725389 - -
          Affymetrix Rat Gene 1.0 ST Array 10725390 - -
          Affymetrix Rat Gene 1.0 ST Array 10725391 - -
          Affymetrix Rat Gene 1.0 ST Array 10725392 - -
          Affymetrix Rat Gene 1.1 ST Array 10725387 - -
          Rat Genome 230 2.0 Array 1374139_at - -
          Rat Genome 230 2.0 Array 1374139_PM_at - -
          Affymetrix HT Rat Focus Array 1374139_at - -
          Affymetrix HT Rat Focus Array 1374139_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100051376.rn.1 TC0100006538.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051374.rn.1 TC0100006538.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051375.rn.1 TC0100006538.rn.1 RTA TC
          Rat Expression Set 230 1374139_at - -
          Rat Expression Set 230 1374139_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725387 - -
          Rat Genome U34 Set rc_AI071702_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21711768 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368793 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline