Search
Search

Ppp2r5d| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001062510.5 | XP_001062510.4 | 3 - 4 | 900 | 90 | |
| XM_006244570.2 | XP_006244632.1 | 3 - 4 | 903 | 90 | ||
| GenBank mRNA | BC099800.1 | - | 2 - 3 | 192 | 90 | |
Set Membership: |
|