Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01511905_m1
          See other DLK2 GE Assays ›
          Gene Symbol:
          Dlk2
          Gene Name:
          delta-like 2 homolog (Drosophila)
          Gene Aliases:
          DLK-2, Egfl9
          Chromosome Location:
          Chr.9: 17061187 - 17065600 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01511905_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dlk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108202.1 NP_001101672.1 3 - 4 262 103
          XM_006244523.2 XP_006244585.1 3 - 4 229 103
          XM_008766865.1 XP_008765087.1 3 - 4 496 103
          GenBank mRNA BC169084.1 - 2 - 3 677 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001576.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of fat cell differentiation
          negative regulation of Notch signaling pathway
          calcium ion binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P286920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83076 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020621 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P020621 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P688422 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926492 - -
          Affymetrix Rat Gene 1.0 ST Array 10926489 - -
          Affymetrix Rat Gene 1.0 ST Array 10926494 - -
          Affymetrix Rat Gene 1.0 ST Array 10926487 - -
          Affymetrix Rat Gene 1.0 ST Array 10926486 - -
          Affymetrix Rat Gene 1.0 ST Array 10926488 - -
          Affymetrix Rat Gene 1.0 ST Array 10926493 - -
          Affymetrix Rat Gene 1.0 ST Array 10926491 - -
          Affymetrix Rat Gene 1.0 ST Array 10926490 - -
          Affymetrix Rat Gene 1.1 ST Array 10926485 - -
          Rat Genome 230 2.0 Array 1384906_at - -
          Rat Genome 230 2.0 Array 1384906_PM_at - -
          Affymetrix HT Rat Focus Array 1384906_PM_at - -
          Affymetrix HT Rat Focus Array 1384906_at - -
          Affymetrix Rat Clariom S Assay PSR0900013137.rn.1 TC0900001576.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013138.rn.1 TC0900001576.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013136.rn.1 TC0900001576.rn.1 RTA TC
          Rat Expression Set 230 1384906_PM_at - -
          Rat Expression Set 230 1384906_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926485 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20762938 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline