Search
Search
| Gene Symbol: Mea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044286.1 | NP_001037751.1 | 2 - 3 | 350 | 62 | |
| XM_006244566.2 | XP_006244628.1 | 1 - 2 | 284 | 62 | ||
| XM_006244567.2 | XP_006244629.1 | 2 - 3 | 405 | 62 | ||
| XM_006244568.2 | XP_006244630.1 | 2 - 3 | 351 | 62 | ||
| XM_006244569.2 | XP_006244631.1 | 2 - 3 | 342 | 62 | ||
| XM_008766880.1 | XP_008765102.1 | 2 - 3 | 450 | 62 | ||
| GenBank mRNA | BC089978.1 | - | 2 - 3 | 350 | 7317693 | 62 |
| FQ213546.1 | - | 2 - 3 | 335 | 62 | ||
| FQ223977.1 | - | 2 - 3 | 336 | 62 | ||
Set Membership: |
|