Search
Search
| Gene Symbol: Itga6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053725.1 | NP_446177.1 | 6 - 7 | 1177 | 53 | |
| XM_006234351.1 | XP_006234413.1 | 6 - 7 | 1374 | 53 | ||
| GenBank mRNA | AJ312934.1 | - | 4 - 5 | 687 | 53 | |
| BF420621.1 | - | 2 - 3 | 283 | 53 | ||
Set Membership: |
|