Search
Search

Ppip5k1| Gene Symbol: Ppip5k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080783.1 | NP_001074252.1 | 22 - 23 | 2774 | 80 | |
| XM_006234838.2 | XP_006234900.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234840.2 | XP_006234902.1 | 23 - 24 | 2968 | 80 | ||
| XM_006234841.2 | XP_006234903.1 | 23 - 24 | 3243 | 80 | ||
| XM_006234843.2 | XP_006234905.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234844.2 | XP_006234906.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234845.2 | XP_006234907.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234848.2 | XP_006234910.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234849.2 | XP_006234911.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234850.2 | XP_006234912.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234851.2 | XP_006234913.1 | 23 - 24 | 2848 | 80 | ||
| XM_006234852.2 | XP_006234914.1 | 23 - 24 | 2848 | 80 | ||
| XM_008762170.1 | XP_008760392.1 | 16 - 17 | 2027 | 80 | ||
Set Membership: |
|