Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01513327_g1
          See other NDUFAF5 GE Assays ›
          Gene Symbol:
          Ndufaf5
          Gene Name:
          NADH dehydrogenase (ubiquinone) complex I, assembly factor 5
          Gene Aliases:
          RGD1309829
          Chromosome Location:
          Chr.3: 133232408 - 133261932 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01513327_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufaf5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001126371.1 NP_001119843.1 9 - 10 901 62
          XM_006235097.2 XP_006235159.1 8 - 9 878 62
          GenBank mRNA BC166551.1 - 9 - 10 901 7307233 62
          FQ229926.1 - 8 - 9 794 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001836.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          methylation
          mitochondrial respiratory chain complex I assembly
          molecular_function
          methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P645352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P645352 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P645352 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P645352 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P645352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840299 - -
          Affymetrix Rat Gene 1.0 ST Array 10840291 - -
          Affymetrix Rat Gene 1.0 ST Array 10840298 - -
          Affymetrix Rat Gene 1.0 ST Array 10840292 - -
          Affymetrix Rat Gene 1.0 ST Array 10840295 - -
          Affymetrix Rat Gene 1.0 ST Array 10840293 - -
          Affymetrix Rat Gene 1.0 ST Array 10840290 - -
          Affymetrix Rat Gene 1.0 ST Array 10840289 - -
          Affymetrix Rat Gene 1.0 ST Array 10840294 - -
          Affymetrix Rat Gene 1.0 ST Array 10840297 - -
          Affymetrix Rat Gene 1.0 ST Array 10840296 - -
          Affymetrix Rat Gene 1.1 ST Array 10840288 - -
          Rat Genome 230 2.0 Array 1373899_at - -
          Rat Genome 230 2.0 Array 1373899_PM_at - -
          Affymetrix HT Rat Focus Array 1373899_at - -
          Affymetrix HT Rat Focus Array 1373899_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300013261.rn.1 TC0300001836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300013260.rn.1 TC0300001836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300013259.rn.1 TC0300001836.rn.1 RTA TC
          Rat Expression Set 230 1373899_at - -
          Rat Expression Set 230 1373899_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840288 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21622985 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline