Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01513815_m1
          See other SPI1 GE Assays ›
          Gene Symbol:
          Spi1
          Gene Name:
          Spi-1 proto-oncogene
          Gene Aliases:
          Pu.1, Sfpi1
          Chromosome Location:
          Chr.3: 79918135 - 79937708 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01513815_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spi1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005892.2 NP_001005892.1 4 - 5 654 62
          XM_006234532.2 XP_006234594.1 4 - 5 1533 62
          GenBank mRNA AB154364.1 - 4 - 5 517 62
          BC091185.1 - 4 - 5 654 7366435 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001127.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spi1

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculature development
          lymphoid progenitor cell differentiation
          myeloid leukocyte differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          lymphocyte differentiation
          cell differentiation
          erythrocyte differentiation
          macrophage differentiation
          granulocyte differentiation
          somatic stem cell population maintenance
          myeloid dendritic cell differentiation
          histone H3 acetylation
          hypermethylation of CpG island
          negative regulation of MHC class II biosynthetic process
          regulation of erythrocyte differentiation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          anatomical structure regression
          cellular response to ethanol
          negative regulation of histone H4 acetylation
          apoptotic process involved in patterning of blood vessels
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          sequence-specific DNA binding
          NFAT protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P433783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62204 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P433783 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P433783 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P433783 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P433783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10837798 - -
          Affymetrix Rat Gene 1.0 ST Array 10837795 - -
          Affymetrix Rat Gene 1.0 ST Array 10837796 - -
          Affymetrix Rat Gene 1.0 ST Array 10837800 - -
          Affymetrix Rat Gene 1.0 ST Array 10837797 - -
          Affymetrix Rat Gene 1.0 ST Array 10837799 - -
          Affymetrix Rat Gene 1.1 ST Array 10837794 - -
          Affymetrix Rat Clariom S Assay PSR0300007875.rn.1 TC0300001127.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10837794 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21902112 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651146 - -
          RatRef-12 Expression BeadChip ILMN_54334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline