Search
Search
| Gene Symbol: Snap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022689.2 | NP_073180.1 | 2 - 3 | 209 | 67 | |
| GenBank mRNA | AF052596.1 | - | 61 | 67 | ||
| BC127494.1 | - | 2 - 3 | 209 | 8361815 | 67 | |
Set Membership: |
|