Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01514778_g1
          See other SIGLEC1 GE Assays ›
          Gene Symbol:
          Siglec1
          Gene Name:
          sialic acid binding Ig-like lectin 1, sialoadhesin
          Gene Aliases:
          Sn
          Chromosome Location:
          Chr.3: 123612067 - 123631845 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01514778_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Siglec1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107777.1 NP_001101247.1 19 - 20 5513 92
          XM_006235024.2 XP_006235086.1 20 - 21 6687 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004191.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P686879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P686879 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P686879 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P686879 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40637 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10850034 - -
          Affymetrix Rat Gene 1.0 ST Array 10850036 - -
          Affymetrix Rat Gene 1.0 ST Array 10850031 - -
          Affymetrix Rat Gene 1.0 ST Array 10850029 - -
          Affymetrix Rat Gene 1.0 ST Array 10850044 - -
          Affymetrix Rat Gene 1.0 ST Array 10850037 - -
          Affymetrix Rat Gene 1.0 ST Array 10850043 - -
          Affymetrix Rat Gene 1.0 ST Array 10850035 - -
          Affymetrix Rat Gene 1.0 ST Array 10850033 - -
          Affymetrix Rat Gene 1.0 ST Array 10850027 - -
          Affymetrix Rat Gene 1.0 ST Array 10850032 - -
          Affymetrix Rat Gene 1.0 ST Array 10850030 - -
          Affymetrix Rat Gene 1.0 ST Array 10850041 - -
          Affymetrix Rat Gene 1.0 ST Array 10850038 - -
          Affymetrix Rat Gene 1.0 ST Array 10850045 - -
          Affymetrix Rat Gene 1.0 ST Array 10850028 - -
          Affymetrix Rat Gene 1.0 ST Array 10850039 - -
          Affymetrix Rat Gene 1.0 ST Array 10850046 - -
          Affymetrix Rat Gene 1.0 ST Array 10850042 - -
          Affymetrix Rat Gene 1.1 ST Array 10850025 - -
          Affymetrix Rat Clariom S Assay PSR0300032650.rn.1 TC0300004191.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300032648.rn.1 TC0300004191.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10850025 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22146548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline