Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01514895_m1
          See other FBN1 GE Assays ›
          Gene Symbol:
          Fbn1
          Gene Name:
          fibrillin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 117569708 - 117766160 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01514895_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031825.1 NP_114013.1 34 - 35 4332 58
          XM_006234935.2 XP_006234997.1 35 - 36 4621 58
          XM_008762202.1 XP_008760424.1 34 - 35 4522 58
          XM_008762203.1 XP_008760425.1 35 - 36 4616 58
          GenBank mRNA AF135059.1 - 34 - 35 4332 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004073.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbn1

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          metanephros development
          kidney development
          heart development
          sequestering of BMP in extracellular matrix
          sequestering of TGFbeta in extracellular matrix
          cellular response to transforming growth factor beta stimulus
          cellular response to insulin-like growth factor stimulus
          extracellular matrix structural constituent
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P179745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31892 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89631 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P179745 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P179745 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P179745 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P996061 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179745 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701905484 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700921102 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700591422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74747 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4733 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95143 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97948 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85923 - -
          Agilent Rat cDNA Microarray (G4105A) 701905484 - -
          Agilent Rat cDNA Microarray (G4105A) 700921102 - -
          Agilent Rat cDNA Microarray (G4105A) 700591422 - -
          Agilent Rat cDNA Microarray (G4105A) 700148655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10849350 - -
          Affymetrix Rat Gene 1.0 ST Array 10849342 - -
          Affymetrix Rat Gene 1.0 ST Array 10849347 - -
          Affymetrix Rat Gene 1.0 ST Array 10849371 - -
          Affymetrix Rat Gene 1.0 ST Array 10849393 - -
          Affymetrix Rat Gene 1.0 ST Array 10849386 - -
          Affymetrix Rat Gene 1.0 ST Array 10849368 - -
          Affymetrix Rat Gene 1.0 ST Array 10849338 - -
          Affymetrix Rat Gene 1.0 ST Array 10849340 - -
          Affymetrix Rat Gene 1.0 ST Array 10849369 - -
          Affymetrix Rat Gene 1.0 ST Array 10849334 - -
          Affymetrix Rat Gene 1.0 ST Array 10849366 - -
          Affymetrix Rat Gene 1.0 ST Array 10849353 - -
          Affymetrix Rat Gene 1.0 ST Array 10849373 - -
          Affymetrix Rat Gene 1.0 ST Array 10849364 - -
          Affymetrix Rat Gene 1.0 ST Array 10849365 - -
          Affymetrix Rat Gene 1.0 ST Array 10849389 - -
          Affymetrix Rat Gene 1.0 ST Array 10849329 - -
          Affymetrix Rat Gene 1.0 ST Array 10849355 - -
          Affymetrix Rat Gene 1.0 ST Array 10849345 - -
          Affymetrix Rat Gene 1.0 ST Array 10849337 - -
          Affymetrix Rat Gene 1.0 ST Array 10849359 - -
          Affymetrix Rat Gene 1.0 ST Array 10849363 - -
          Affymetrix Rat Gene 1.0 ST Array 10849379 - -
          Affymetrix Rat Gene 1.0 ST Array 10849352 - -
          Affymetrix Rat Gene 1.0 ST Array 10849383 - -
          Affymetrix Rat Gene 1.0 ST Array 10849370 - -
          Affymetrix Rat Gene 1.0 ST Array 10849382 - -
          Affymetrix Rat Gene 1.0 ST Array 10849332 - -
          Affymetrix Rat Gene 1.0 ST Array 10849335 - -
          Affymetrix Rat Gene 1.0 ST Array 10849390 - -
          Affymetrix Rat Gene 1.0 ST Array 10849356 - -
          Affymetrix Rat Gene 1.0 ST Array 10849377 - -
          Affymetrix Rat Gene 1.0 ST Array 10849339 - -
          Affymetrix Rat Gene 1.0 ST Array 10849372 - -
          Affymetrix Rat Gene 1.0 ST Array 10849375 - -
          Affymetrix Rat Gene 1.0 ST Array 10849392 - -
          Affymetrix Rat Gene 1.0 ST Array 10849380 - -
          Affymetrix Rat Gene 1.0 ST Array 10849358 - -
          Affymetrix Rat Gene 1.0 ST Array 10849344 - -
          Affymetrix Rat Gene 1.0 ST Array 10849388 - -
          Affymetrix Rat Gene 1.0 ST Array 10849341 - -
          Affymetrix Rat Gene 1.0 ST Array 10849351 - -
          Affymetrix Rat Gene 1.0 ST Array 10849330 - -
          Affymetrix Rat Gene 1.0 ST Array 10849333 - -
          Affymetrix Rat Gene 1.0 ST Array 10849336 - -
          Affymetrix Rat Gene 1.0 ST Array 10849362 - -
          Affymetrix Rat Gene 1.0 ST Array 10849361 - -
          Affymetrix Rat Gene 1.0 ST Array 10849376 - -
          Affymetrix Rat Gene 1.0 ST Array 10849328 - -
          Affymetrix Rat Gene 1.0 ST Array 10849346 - -
          Affymetrix Rat Gene 1.0 ST Array 10849354 - -
          Affymetrix Rat Gene 1.0 ST Array 10849357 - -
          Affymetrix Rat Gene 1.0 ST Array 10849384 - -
          Affymetrix Rat Gene 1.0 ST Array 10849331 - -
          Affymetrix Rat Gene 1.0 ST Array 10849343 - -
          Affymetrix Rat Gene 1.0 ST Array 10849348 - -
          Affymetrix Rat Gene 1.0 ST Array 10849374 - -
          Affymetrix Rat Gene 1.1 ST Array 10849327 - -
          Rat Genome 230 2.0 Array 1387351_at - -
          Rat Genome 230 2.0 Array 1368829_at - -
          Rat Genome 230 2.0 Array 1368829_PM_at - -
          Rat Genome 230 2.0 Array 1387351_PM_at - -
          Affymetrix HT Rat Focus Array 1368829_PM_at - -
          Affymetrix HT Rat Focus Array 1368829_at - -
          Affymetrix HT Rat Focus Array 1387351_at - -
          Affymetrix HT Rat Focus Array 1387351_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300031408.rn.1 TC0300004073.rn.1 RTA TC
          Rat Expression Set 230 1387351_PM_at - -
          Rat Expression Set 230 1368829_PM_at - -
          Rat Expression Set 230 1368829_at - -
          Rat Expression Set 230 1387351_at - -
          Affymetrix Rat Gene 1.0 ST Array 10849327 - -
          Rat Genome U34 Set rc_AA997765_at - -
          Rat Genome U34 Set rc_AI230154_at - -
          Rat Genome U34 Set rc_AA859321_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21452608 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359541 - -
          RatRef-12 Expression BeadChip ILMN_51879 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline