Search
Search

Slc12a6| Gene Symbol: Slc12a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109630.1 | NP_001103100.1 | 8 - 9 | 1117 | 59 | |
| XM_006234710.2 | XP_006234772.1 | 7 - 8 | 2092 | 59 | ||
| XM_006234711.2 | XP_006234773.1 | 8 - 9 | 1674 | 59 | ||
Set Membership: |
|