Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515170_m1
          See other PIH1D3 GE Assays ›
          Gene Symbol:
          Pih1d3
          Gene Name:
          PIH1 domain containing 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 111389557 - 111439399 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515170_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pih1d3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024890.1 NP_001020061.1 6 - 7 555 73
          GenBank mRNA BC097458.1 - 6 - 7 555 7457221 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000950.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pih1d3

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P078403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79360 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P078403 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P078403 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P459626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80148 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45996 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20004 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935185 - -
          Affymetrix Rat Gene 1.0 ST Array 10935186 - -
          Affymetrix Rat Gene 1.0 ST Array 10935181 - -
          Affymetrix Rat Gene 1.0 ST Array 10935180 - -
          Affymetrix Rat Gene 1.0 ST Array 10935187 - -
          Affymetrix Rat Gene 1.0 ST Array 10935182 - -
          Affymetrix Rat Gene 1.0 ST Array 10935183 - -
          Affymetrix Rat Gene 1.0 ST Array 10935184 - -
          Affymetrix Rat Gene 1.1 ST Array 10935179 - -
          Affymetrix Rat Clariom S Assay PSR0X00005815.rn.1 TC0X00000950.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00005814.rn.1 TC0X00000950.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10935179 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21335993 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372878 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline